Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Otubains are deubiquitylating cysteine proteases that belong to the ovarian tumor protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin -protein bond.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme OTUB2; FLJ21916; MGC3102; OTU domain, ubiquitin aldehyde binding 2; OTU domain-containing ubiquitin aldehyde-binding protein 2; Otubain-2; Ubiquitin thioesterase OTUB2; ubiquitin-specific protease otubain 2; Ubiquitin-specific-processing protease OTUB2
Gene Aliases: C14orf137; OTB2; OTU2; OTUB2
UniProt ID: (Human) Q96DC9
Entrez Gene ID: (Dog) 480420, (Human) 78990, (Rat) 314405
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support