Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: Epithelial cells in normal placenta or prostate carcinoma.
MA1-25811 is expected to cross react with all known eukaryotic ODC proteins due to sequence homology.
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC103389; ODC; Ornithine decarboxylase; Ornitine decarboxylase
Gene Aliases: ODC; ODC1
UniProt ID: (Human) P11926, (Rat) P09057, (Mouse) P00860
Entrez Gene ID: (Human) 4953, (Rat) 24609, (Mouse) 18263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support