Search Thermo Fisher Scientific
PA1-20590 detects OA1 in Human samples.
PA1-20590 has been successfully used in Immunohistochemistry (Paraffin) procedures.
PA1-20590 immunogen corresponds to Synthetic peptide (aa sequence is considered to be commercially sensitive).