Search Thermo Fisher Scientific
PA1-10882 detects SOX6 in human and mouse samples. This antibody is predicted to recognize rat and chimapanzee SOX6 based on 100% sequence homology.
PA1-10882 has been successfully used in Western blot procedures.
PA1-10882 immunogen is a synthetic peptide corresponding to residues 780-796 of human and mouse SOX6.
Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.
Protein Aliases: SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box containing gene 6; Transcription factor SOX-6
Gene Aliases: AI987981; HSSOX6; Sox-6; SOX-LZ; SOX6; SOXD
UniProt ID: (Human) P35712, (Mouse) P40645
Entrez Gene ID: (Human) 55553, (Mouse) 20679