Invitrogen
PA1-74362 detects Rabbit IgA from rabbit samples and shows no cross-reactivity with other rabbit IG classes in immunoelectrophoresis.
Goat anti Rabbit IgA antibody recognizes rabbit IgA and shows no cross-reactivity with other rabbit immunoglobulin classes in immunoelectrophoresis.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.