Search Thermo Fisher Scientific
A suggested positive control is mouse brain tissue lysate.
PA5-20476 can be used with blocking peptide PEP-0596.
SLIT and NTRK-like family 4 (Slitrk4) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk4 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
Protein Aliases: SLIT and NTRK-like family, member 4; SLIT and NTRK-like protein 4; slit and trk like gene 4
Gene Aliases: 6430513G13; D930039I09Rik; SLITRK4
UniProt ID: (Human) Q96DL3, (Mouse) Q810B8
Entrez Gene ID: (Human) 139065, (Mouse) 245446, (Rat) 302473