Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (94%), Rat (94%), Xenopus laevis (83%), Rhesus Monkey (98%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Protein Aliases: brix domain containing 1; Brix domain-containing protein 1; homolog of Rpf2; ribosomal processing factor 2 homolog; Ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; RP11-397G5.1
Gene Aliases: bA397G5.4; BXDC1; RPF2
UniProt ID: (Human) Q9H7B2
Entrez Gene ID: (Human) 84154