Search Thermo Fisher Scientific
Immunogen sequence: PISKDDVERC KQKDLLEQMM AEMIGEFPDL HRTIVSERDV YLTYMLRQAA RRLELPRASD AEPRKCVPSV VVGVVGMGHV PGIEKNWSTD LNIQEIMTVP PPSVSGRVSR LAVKAAFFGL LGYSLYWMGR RTASLVLSLP AAQYCLQR
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
The function of TRABD remains unknown.