Search Thermo Fisher Scientific
Immunogen sequence: VFSNTQRFLS QHRCGEPEAS PPRTLL
Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 69%.
The function of this protein remains unknown.