Search Thermo Fisher Scientific
Predicted to react with Mouse samples.
Regulates cell motility and proliferation. May have phosphatase activity. Reduces AKT1 phosphorylation. Lowers AKT1 kinase activity and interferes with AKT1 signaling.