Search Thermo Fisher Scientific
Immunogen sequence: TSCLGSTSSP FVIFPEIVRN GDPSTWVKNS TALISTIPGT YVGVANPVPA SLLLNKDPNL GLNRDPRHLP KQEPISIID
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The protein encoded by this gene is a transcriptional corepressor that is found tethered to promoter regions by DNA-binding proteins. The encoded protein can interact with several different class II histone deacetylases to repress transcription. Two transcript variants encoding different isoforms have been found for this gene.