Search Thermo Fisher Scientific
Immunogen sequence: PDSDDEKIRL LLRKHRAAFS VLSLGA
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The NKX family of homeodomain proteins controls numerous developmental processes. Members of the NKX6 subfamily, including NKX6-3, are involved in development of the central nervous system (CNS), gastrointestinal tract, and pancreas.