Search Thermo Fisher Scientific
Immunogen sequence: LKSQGKNCKK LAKNLLKEPE NCNNFCLWKQ YAHLEWLLGN TEDARKVFDT ALGMAGSREL KDSDLCELSL LYAELEVELS PEVRRAATAR AVHILTKLTE SSPYGPYTGQ VLAVHILKAR KAYEHALQDC LGDSCVSNPA
Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
NRDE2 gene ontology (GO) annotations include RNA interference; chromatin silencing by small RNA.