Search Thermo Fisher Scientific
Immunogen sequence: LFVKVKLTAL GCSRIAIKKY LRAGYD
Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 35%.
CLIC6 is a member of the chloride intracellular channel family of proteins. The gene is part of a large triplicated region found on chromosomes 1, 6, and 21. An alternatively spliced transcript variant has been described, but its biological validity has not been determined.