Search Thermo Fisher Scientific
Synthetic peptide sequence: 653-671aa, YTIRQAQLQDAGIYECESK.
Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.