Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
MA1-34079 detects PABP from human, chicken, rabbit and Xenopus laevis samples. Does not react wtih Drosophilia melanogaster or mouse samples.
The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic poly(A)-binding protein 1-A; PABP-1; PABP-1-A; PABP-A; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1-A; Polyadenylate-binding protein 1; xPABP1-A
Gene Aliases: PAB1; PABP; PABP1; PABPC1; pabpc1-a; pabpc1-b; pabpc1.L; PABPC2; PABPL1; XELAEV_18033915mg
UniProt ID: (Human) P11940
Entrez Gene ID: (Human) 26986, (Chicken) 430997, (Rabbit) 100342434, (Xenopus) 379896
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support