Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
Immunogen sequence: TWHSLRNSPG EKINLLKRSL LSIPNTDYIQ LLSEIAKEQG FNITYLDIDE LSANGQYQCL AELSTSPITV CHGSGISCGN AQSDAAHNAL QYLKIIAERK
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interferon-inducible double stranded RNA-dependent protein kinase activator A; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PKR associated protein X; PKR-associated protein X; PKR-associating protein X; Protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double stranded RNA dependent activator; protein kinase, interferon-inducible double stranded RNA-dependent activator; Protein kinase, interferon-inducible double-stranded RNA-dependent activator; RAX
Gene Aliases: AV120107; DYT16; HSD-14; HSD14; lear; PACT; PRK; PRKRA; RAX
UniProt ID: (Human) O75569, (Rat) Q4V8C7, (Mouse) Q9WTX2
Entrez Gene ID: (Human) 8575, (Rat) 311130, (Mouse) 23992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support