Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with bovine, mouse, rat, rabbit and Xenopus based on sequence homology.
PAK1 is a serine/threonine kinase that is a target for restenosis, cancer, and immunosuppression treatment. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology.