Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Human squamous cell lung carcinoma tissue
Cellular location: Secreted.
The platelet-derived growth factor (PDGF) family of disulfide-linked dimeric proteins consists of homo/hetero dimeric proteins, PDGF-AA, -BB, -CC, -DD and -AB. The PDGF-AA, -BB and AB bind to alpha type receptor, while beta type receptors bind to PDGF-BB and PDGF-AB. Upon activation by PDGF, the receptors get dimerised and auphosphorylated in several sites on its receptors (Receptor tyrosine kinase) in cytosolic domain to trigger cell signaling through PI3K pathway. All PDGF isoforms are synthesized as inactive precursors and are processed to active forms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: becaplermin; c-sis; PDGF A-chain; pdgf protein; PDGF subunit A; PDGF subunit B; PDGF, B chain; PDGF-1; PDGF-2; platelet derived growth factor, alpha; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; Platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; Platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit A; Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); Proto-oncogene c-Sis
Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF-A; PDGF-B; PDGF1; PDGF2; PDGFA; PDGFACP; PDGFB; Rpa1; SIS; SSV
UniProt ID: (Human) P04085, (Human) P01127, (Rat) P28576, (Mouse) P20033, (Mouse) P31240, (Rat) Q05028
Entrez Gene ID: (Human) 5154, (Human) 5155, (Rat) 25266, (Mouse) 18590, (Mouse) 18591, (Rat) 24628
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support