Search Thermo Fisher Scientific
This antibody is predicted to react with bovine based on sequence homology.
PEX11A may be involved in peroxisomal proliferation and may regulate peroxisomes division. It may mediate binding of coatomer proteins to the peroxisomal membrane.