Product References
BLOC-1 and BLOC-3 regulate VAMP7 cycling to and from melanosomes via distinct tubular transport carriers.
The Journal of cell biology
Dennis MK,Delevoye C,Acosta-Ruiz A,Hurbain I,Romao M,Hesketh GG,Goff PS,Sviderskaya EV,Bennett DC,Luzio JP,Galli T,Owen DJ,Raposo G,Marks MS
MA5-16712 was used in immunocytochemistry to propose that VAMP7 mediates fusion of BLOC-1-dependent transport carriers with melanosomes
Mon Aug 01 00:00:00 EDT 2016
Retroperitoneal lymphangioma: A report of 2 cases and a review of the literature regarding the differential diagnoses of retroperitoneal cystic masses.
Oncology letters
DI Marco M,Grassi E,Vecchiarelli S,Durante S,Macchini M,Biasco G
MA5-16712 was used in immunohistochemistry - paraffin section to review 2 cases of retroperitoneal lymphangioma and literature regarding the differential diagnoses of retroperitoneal cystic masses
Sun May 01 00:00:00 EDT 2016
NK cell activating receptor ligand expression in lymphangioleiomyomatosis is associated with lung function decline.
JCI insight
Osterburg AR,Nelson RL,Yaniv BZ,Foot R,Donica WR,Nashu MA,Liu H,Wikenheiser-Brokamp KA,Moss J,Gupta N,McCormack FX,Borchers MT
MA516712 was used in immunohistochemistry - frozen section to investigate the role of natural killer cells to lymphangioleiomyomatosis pathogenesis
Thu Oct 06 00:00:00 EDT 2016
BLOC-2 targets recycling endosomal tubules to melanosomes for cargo delivery.
The Journal of cell biology
Dennis MK,Mantegazza AR,Snir OL,Tenza D,Acosta-Ruiz A,Delevoye C,Zorger R,Sitaram A,de Jesus-Rojas W,Ravichandran K,Rux J,Sviderskaya EV,Bennett DC,Raposo G,Marks MS,Setty SR
MA5-16712 was used in flow cytometry to investigate the role of BLOC-2 in endosomal tubular transport in melanosomes
Mon May 25 00:00:00 EDT 2015
ASPL-TFE3 translocation in vulvovaginal alveolar soft part sarcoma.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Jabbour MN,Seoud M,Al-Ahmadie H,Abdul-Karim FW,Zaatari GS
MA5-16712 was used in immunohistochemistry to report on a case of vulvovaginal alveolar soft part sarcoma
Thu May 01 00:00:00 EDT 2014
Targeting BRAFV600E in an inducible murine model of melanoma.
The American journal of pathology
Hooijkaas AI,Gadiot J,van der Valk M,Mooi WJ,Blank CU
MA5-16712 was used in immunohistochemistry to study the effects of targeting BRAF(V600E) on melanoma growth in an inducible murine model
Sat Sep 01 00:00:00 EDT 2012
Differential recognition of a dileucine-based sorting signal by AP-1 and AP-3 reveals a requirement for both BLOC-1 and AP-3 in delivery of OCA2 to melanosomes.
Molecular biology of the cell
Sitaram A,Dennis MK,Chaudhuri R,De Jesus-Rojas W,Tenza D,Setty SR,Wood CS,Sviderskaya EV,Bennett DC,Raposo G,Bonifacino JS,Marks MS
MA5-16712 was used in immunocytochemistry to study the delivery of OCA2 to melanosomes and the distinct roles played by AP-1 and AP-3
Wed Aug 01 00:00:00 EDT 2012
Effect of PUVA therapy on melanocytes and keratinocytes in non-segmental vitiligo: histopathological, immuno-histochemical and ultrastructural study.
Photodermatology, photoimmunology & photomedicine
Anbar TS,El-Sawy AE,Attia SK,Barakat MT,Moftah NH,El-Ammawy TS,Abdel-Rahman AT,El-Tonsy MH
MA5-16712 was used in immunohistochemistry to study the effects of PUVA treatment on melanocytes and keratinocytes from patients with non-segmental vitiligo
Wed Feb 01 00:00:00 EST 2012
Sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression: report of a case and literature review.
Diagnostic pathology
Yin WH,Li J,Chan JK
MA5-16712 was used in immunohistochemistry to report on a case of sporadic haemangioblastoma of the kidney with rhabdoid features and focal CD10 expression
Thu Apr 12 00:00:00 EDT 2012
Cardiac Epithelioid PEComa: Report of Two Cases and Review of the Literature.
Case reports in medicine
Niu H,Wang FW,Zhang PJ,Bing Z
MA5-16712 was used in immunohistochemistry - paraffin section to present two cases of epithelioid PEComas
Fri Sep 14 00:00:00 EDT 2012
Proprotein convertases process Pmel17 during secretion.
The Journal of biological chemistry
Leonhardt RM,Vigneron N,Rahner C,Cresswell P
MA5-16712 was used in immunocytochemistry and western blot to study Pmel17 processing in melanocytes
Fri Mar 18 00:00:00 EDT 2011
BLOC-1 is required for cargo-specific sorting from vacuolar early endosomes toward lysosome-related organelles.
Molecular biology of the cell
Setty SR,Tenza D,Truschel ST,Chou E,Sviderskaya EV,Theos AC,Lamoreux ML,Di Pietro SM,Starcevic M,Bennett DC,Dell'Angelica EC,Raposo G,Marks MS
MA5-16712 was used in immunocytochemistry to investigate the role of BLOC-1 in melanosome maturation
Thu Mar 01 00:00:00 EST 2007