Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GGLLAFAKRR NCCIGCRTVL SHQGAVCEFC QPRESELYQK EVSHLNALEE RFSRLWTQCQ RCQGSLHEDV ICTSRDCPIF YMRKKVRKDL EDQEQLLRRF GPPGPEAW
This gene encodes the 125-kDa catalytic subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3' to 5' exonuclease activity and plays a critical role in DNA replication and repair. Alternatively spliced transcript variants have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' exodeoxyribonuclease; CDC2 homolog; DNA polymerase delta 1, catalytic domain; DNA polymerase delta catalytic subunit; DNA polymerase delta, catalytic subunit; DNA polymerase subunit delta p125; DNA-directed DNA polymerase delta 1; POL D; polymerase (DNA directed), delta 1, catalytic subunit 125kDa; polymerase (DNA) delta 1, catalytic subunit; Polymerase D
Gene Aliases: 125kDa; CDC2; CRCS10; MDPL; POLD; POLD1
UniProt ID: (Human) P28340, (Rat) O54747, (Mouse) P52431
Entrez Gene ID: (Human) 5424, (Rat) 59294, (Mouse) 18971
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support