Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes the fifth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit is shared by the other two DNA-directed RNA polymerases and is present in two-fold molar excess over the other polymerase subunits. An interaction between this subunit and a hepatitis virus transactivating protein has been demonstrated, suggesting that interaction between transcriptional activators and the polymerase can occur through this subunit. A pseudogene is located on chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA directed RNA polymerase II 23 kda polypeptide; DNA-directed RNA polymerase II 23 kDa polypeptide; DNA-directed RNA polymerase II subunit E; DNA-directed RNA polymerase subunit RPABC1; DNA-directed RNA polymerases I, II, and III subunit RPABC1; polymerase (RNA) II (DNA directed) polypeptide E, 25kDa; polymerase (RNA) II subunit E; RNA polymerase II subunit E; RNA polymerases I, II, and III subunit ABC1; RPB5 homolog; XAP4
Gene Aliases: 2410021N14Rik; 25kDa; AW208866; hRPB25; hsRPB5; POLR2E; RPABC1; RPB5; XAP4
UniProt ID: (Human) P19388, (Mouse) Q80UW8, (Rat) B0BNE2
Entrez Gene ID: (Dog) 611859, (Human) 5434, (Mouse) 66420, (Rat) 690966
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support