Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase II 13.3 kDa polypeptide; DNA-directed RNA polymerase II subunit J; DNA-directed RNA polymerase II subunit J-1; DNA-directed RNA polymerase II subunit RPB11; DNA-directed RNA polymerase II subunit RPB11-a; polymerase (RNA) II (DNA directed) polypeptide I; polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa; polymerase (RNA) II subunit J; RNA polymerase II 13.3 kDa subunit; RNA polymerase II 4 (14 kDa subunit); RNA polymerase II subunit B11; RNA polymerase II subunit B11-a; RNA polymerase II subunit RPB14; RPB14
Gene Aliases: 14.5kDa; hRPB14; Polr2i; POLR2J; POLR2J1; RPB11; RPB11A; RPB11m; Rpo2-4
UniProt ID: (Human) P52435
Entrez Gene ID: (Human) 5439, (Rat) 288588, (Mouse) 20022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support