Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINH
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. Also a component of RNase MRP complex, which cleaves pre-rRNA sequences.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPOP7; POP7 (processing of precursor, S. cerevisiae) homolog; processing of precursor 7, ribonuclease P subunit; processing of precursor 7, ribonuclease P/MRP subunit; Ribonuclease P protein subunit p20; Ribonucleases P/MRP protein subunit POP7 homolog; RNaseP protein p20
Gene Aliases: 0610037N12Rik; POP7; RPP2; RPP20
UniProt ID: (Human) O75817
Entrez Gene ID: (Human) 10248
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support