Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Mitochondrial RNA-binding protein that has a role in mitochondrial translation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S39, mitochondrial; MRP-S39; Pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Small ribosomal subunit protein mS39; Transformation-related gene 15 protein; TRG-15
Gene Aliases: MRP-S39; MRPS39; PTCD3; TRG15
UniProt ID: (Human) Q96EY7
Entrez Gene ID: (Human) 55037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support