Product References
Label2label: training a neural network to selectively restore cellular structures in fluorescence microscopy.
Journal of cell science
Kölln LS,Salem O,Valli J,Hansen CG,McConnell G
Published figure using Paxillin monoclonal antibody (Product # MA5-13356) in Immunocytochemistry
Tue Feb 01 00:00:00 EST 2022
Label2label: training a neural network to selectively restore cellular structures in fluorescence microscopy.
Journal of cell science
Kölln LS,Salem O,Valli J,Hansen CG,McConnell G
Published figure using Paxillin monoclonal antibody (Product # MA5-13356) in Immunocytochemistry
Tue Feb 01 00:00:00 EST 2022
Fibroblast growth factor 2 inhibits myofibroblastic activation of valvular interstitial cells.
PloS one
Ground M,Waqanivavalagi S,Park YE,Callon K,Walker R,Milsom P,Cornish J
MA5-13356 was used in Immunocytochemistry to show that FGF-2 can reverse the myofibroblastic phenotype of VICs isolated from diseased valves and inhibit the calcification potential of these cells.
Tue Jun 21 00:00:00 EDT 2022
Topography-induced large-scale antiparallel collective migration in vascular endothelium.
Nature communications
Leclech C,Gonzalez-Rodriguez D,Villedieu A,Lok T,Déplanche AM,Barakat AI
MA5-13356 was used in Flow Cytometry to posit that in unconfined cell assemblies, physical factors that constrain or bias cellular orientation such as anisotropic extracellular matrix cues or directed flow-derived shear forces dictate the pattern of collective cell movement.
Thu May 19 00:00:00 EDT 2022
Coordination of mitochondrial and cellular dynamics by the actin-based motor Myo19.
Journal of cell science
Majstrowicz K,Honnert U,Nikolaus P,Schwarz V,Oeding SJ,Hemkemeyer SA,Bähler M
MA5-13356 was used in Western Blot to investigate which mitochondrial and cellular processes depend on the coordination of Myo19 and microtubule-based motor activities.
Sat May 15 00:00:00 EDT 2021
Coordination of mitochondrial and cellular dynamics by the actin-based motor Myo19.
Journal of cell science
Majstrowicz K,Honnert U,Nikolaus P,Schwarz V,Oeding SJ,Hemkemeyer SA,Bähler M
MA5-13356 was used in Western Blot to investigate which mitochondrial and cellular processes depend on the coordination of Myo19 and microtubule-based motor activities.
Sat May 15 00:00:00 EDT 2021
Carbon Nanotube-Hydrogel Composites Facilitate Neuronal Differentiation While Maintaining Homeostasis of Network Activity.
Advanced materials (Deerfield Beach, Fla.)
Ye L,Ji H,Liu J,Tu CH,Kappl M,Koynov K,Vogt J,Butt HJ
MA5-13356 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to indicate that the composites are promising materials that could indeed facilitate neuronal differentiation while maintaining neuronal homeostasis..
Fri Oct 01 00:00:00 EDT 2021
Roles of Endocytic Processes and Early Endosomes on Focal Adhesion Dynamics in MDA-MB-231 Cells.
Reports of biochemistry & molecular biology
Alharbi BF,Al-Fahad D,Richard Dash P
MA5-13356 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to suggest that there is a link between FA and early endosome markers, which indicates that the early endosomes may be involved in FA dynamics.
Thu Jul 01 00:00:00 EDT 2021
HDAC6 regulates microtubule stability and clustering of AChRs at neuromuscular junctions.
The Journal of cell biology
Osseni A,Ravel-Chapuis A,Thomas JL,Gache V,Schaeffer L,Jasmin BJ
MA5-13356 was used in Western Blotting to show HDAC6 preferentially accumulates at neuromuscular junctions (NMJs) and that it contributes to the organization and the stability of NMJs.
Mon Aug 03 00:00:00 EDT 2020
HDAC6 regulates microtubule stability and clustering of AChRs at neuromuscular junctions.
The Journal of cell biology
Osseni A,Ravel-Chapuis A,Thomas JL,Gache V,Schaeffer L,Jasmin BJ
MA5-13356 was used in Western Blotting to show HDAC6 preferentially accumulates at neuromuscular junctions (NMJs) and that it contributes to the organization and the stability of NMJs.
Mon Aug 03 00:00:00 EDT 2020
HDAC6 regulates microtubule stability and clustering of AChRs at neuromuscular junctions.
The Journal of cell biology
Osseni A,Ravel-Chapuis A,Thomas JL,Gache V,Schaeffer L,Jasmin BJ
MA5-13356 was used in Western Blotting to show HDAC6 preferentially accumulates at neuromuscular junctions (NMJs) and that it contributes to the organization and the stability of NMJs.
Mon Aug 03 00:00:00 EDT 2020
GPR124 facilitates pericyte polarization and migration by regulating the formation of filopodia during ischemic injury.
Theranostics
Chen DY,Sun NH,Lu YP,Hong LJ,Cui TT,Wang CK,Chen XH,Wang SS,Feng LL,Shi WX,Fukunaga K,Chen Z,Lu YM,Han F
Published figure using Paxillin monoclonal antibody (Product # MA5-13356) in Immunofluorescence
Mon Aug 31 00:00:00 EDT 2020
GPR124 facilitates pericyte polarization and migration by regulating the formation of filopodia during ischemic injury.
Theranostics
Chen DY,Sun NH,Lu YP,Hong LJ,Cui TT,Wang CK,Chen XH,Wang SS,Feng LL,Shi WX,Fukunaga K,Chen Z,Lu YM,Han F
Published figure using Paxillin monoclonal antibody (Product # MA5-13356) in Immunofluorescence
Mon Aug 31 00:00:00 EDT 2020
GPR124 facilitates pericyte polarization and migration by regulating the formation of filopodia during ischemic injury.
Theranostics
Chen DY,Sun NH,Lu YP,Hong LJ,Cui TT,Wang CK,Chen XH,Wang SS,Feng LL,Shi WX,Fukunaga K,Chen Z,Lu YM,Han F
Published figure using Paxillin monoclonal antibody (Product # MA5-13356) in Immunofluorescence
Mon Aug 31 00:00:00 EDT 2020
Targeting focal adhesion turnover in invasive breast cancer cells by the purine derivative reversine.
British journal of cancer
Bijian K,Lougheed C,Su J,Xu B,Yu H,Wu JH,Riccio K,Alaoui-Jamali MA
MA5-13356 was used in western blot to study the beneficial effects of reversine-mediated disruption of focal adhesion turnover on breast cancer metastasis
Tue Nov 26 00:00:00 EST 2013
Decreased ezrin and paxillin expression in human urothelial bladder tumors correlate with tumor progression.
Urologic oncology
Athanasopoulou A,Aroukatos P,Nakas D,Repanti M,Papadaki H,Bravou V
MA5-13356 was used in immunohistochemistry to study the clinical significance of ezrin and paxillin downregulation in human urothelial bladder tumors
Thu Aug 01 00:00:00 EDT 2013
Expression of claudin, paxillin and FRA-1 in non-nodular breast lesions in association with microcalcifications.
Sao Paulo medical journal = Revista paulista de medicina
Kandelman JD,Waitzberg AF,Szejnfeld J,Smith RL
MA5-13356 was used in immunohistochemistry to study non-nodular breast lesions for the expression of claudin, paxillin and FRA-1
Fri Nov 15 00:00:00 EST 2013
Paxillin expression and amplification in early lung lesions of high-risk patients, lung adenocarcinoma and metastatic disease.
Journal of clinical pathology
Mackinnon AC,Tretiakova M,Henderson L,Mehta RG,Yan BC,Joseph L,Krausz T,Husain AN,Reid ME,Salgia R
MA5-13356 was used in immunohistochemistry to study paxillin expression and amplification in early lung lesions and its role in lung carcinogenesis
Sat Jan 01 00:00:00 EST 2011
The influence of tubular phenotypic changes on the development of diffuse interstitial fibrosis in renal allografts.
Transplantation proceedings
Özdemir BH,Özdemir AA,Colak T,Sezer S,Haberal M
MA5-13356 was used in immunohistochemistry to study the role of tubular phenotypic changes in the development of diffuse interstitial fibrosis in renal allografts
Tue Mar 01 00:00:00 EST 2011
Extraribosomal function of metallopanstimulin-1: reducing paxillin in head and neck squamous cell carcinoma and inhibiting tumor growth.
International journal of cancer
Dai Y,Pierson SE,Dudney WC,Stack BC
MA5-13356 was used in western blot to investigate the effect of metallopanstimulin-1 on tumor growth and gene expression
Mon Feb 01 00:00:00 EST 2010
Cyclic strain modulates migration and proliferation of vascular smooth muscle cells via Rho-GDIalpha, Rac1, and p38 pathway.
Journal of cellular biochemistry
Qi YX,Qu MJ,Yan ZQ,Zhao D,Jiang XH,Shen BR,Jiang ZL
MA5-13356 was used in immunohistochemistry to study the mechanism for the induction of vascular smooth muscle cell migration and proliferation by cyclic strain
Thu Apr 01 00:00:00 EDT 2010
Focal adhesion plaque associated cytoskeletons are involved in the invasion and metastasis of human colorectal carcinoma.
Cancer investigation
Yang HJ,Chen JZ,Zhang WL,Ding YQ
MA5-13356 was used in immunohistochemistry to investigate the involvement of focal adhesion plaque associated cytoskeletons in the invasion and metastasis of colorectal carcinoma
Mon Feb 01 00:00:00 EST 2010
Vascular smooth muscle cells promote endothelial cell adhesion via microtubule dynamics and activation of paxillin and the extracellular signal-regulated kinase (ERK) pathway in a co-culture system.
European journal of cell biology
Wang YH,Yan ZQ,Shen BR,Zhang L,Zhang P,Jiang ZL
MA5-13356 was used in immunocytochemistry to investigate the importance of vascular smooth muscle cells in endothelial cell adhesion
Sun Nov 01 00:00:00 EDT 2009
Presenilin 1 affects focal adhesion site formation and cell force generation via c-Src transcriptional and posttranslational regulation.
The Journal of biological chemistry
Waschbüsch D,Born S,Niediek V,Kirchgessner N,Tamboli IY,Walter J,Merkel R,Hoffmann B
MA5-13356 was used in western blot to assess the function of presenilins in the cell-matrix interactions of mouse embryonic fibroblasts
Fri Apr 10 00:00:00 EDT 2009
The expression of the cytoskeletal focal adhesion protein paxillin in breast cancer correlates with HER2 overexpression and may help predict response to chemotherapy: a retrospective immunohistochemical study.
The breast journal
Short SM,Yoder BJ,Tarr SM,Prescott NL,Laniauskas S,Coleman KA,Downs-Kelly E,Pettay JD,Choueiri TK,Crowe JP,Tubbs RR,Budd TG,Hicks DG
MA5-13356 was used in immunohistochemistry to investigate the association between paxillin expression and HER2 expression in breast cancer patients
Tue Apr 17 00:00:00 EDT 2007
Smooth muscle alpha-actin deficiency in myofibroblasts leads to enhanced renal tissue fibrosis.
The Journal of biological chemistry
Takeji M,Moriyama T,Oseto S,Kawada N,Hori M,Imai E,Miwa T
MA5-13356 was used in western blot to examine the role of smooth muscle alpha-actin deficiency in renal tissue fibrosis
Fri Dec 29 00:00:00 EST 2006
Sequential activation of RhoA and FAK/paxillin leads to ATP release and actin reorganization in human endothelium.
The Journal of physiology
Hirakawa M,Oike M,Karashima Y,Ito Y
MA5-13356 was used in Western Blotting to investigate the cellular mechanisms of mechanical stress-induced immediate responses in human umbilical vein endothelial cells.
Thu Jul 15 00:00:00 EDT 2004