Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Description: This 2A12 monoclonal antibody reacts with the transcriptional activator PDX1, also known as pancreatic and duodenal homeobox protein and insulin promoter factor. Early expression of PDX1 during development marks the endoderm from which the pancreas, stomach, and duodenum derive. In the mature pancreas, expression is found in insulin-expressing beta-cells. PDX1 functions as an important regulator of several key pancreatic proteins including insulin, somatostatin, glucokinase, islet amyloid factor, and glucose transporter type 2. Deficiency in PDX1 results in a dysregulation of the beta-cell compensation of insulin resistance, which can lead to late-onset type 2 diabetes mellitus (formerly NIDDM) and maturity onset diabetes of the young type 4 (MODY4).
Applications Reported: This 2A12 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.
Applications Tested: This 2A12 antibody has been tested by immunohistochemistry on FFPE human pancreas (with IHC Antigen Retrieval Solution - Low pH (cat. 00-4955)) and can be used at less than or equal to 5 µg/mL. Is is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support