Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Published species |
Fruit fly |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide sequence around phosphorylation site of threonine 305 (I-L-T(p)-T-M) derived from Human CaMK2-alpha-beta-delta |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 50% glycerol, 150mM NaCl |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2554440 |
A suggested positive control for Western blot is NIH-3T3 cells; suggested positive control for IHC is human brain tissue.
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-CaMKII; Calci; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II delta; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase II beta; calcium/calmodulin-dependent protein kinase II delta; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II delta chain; Calcium/calmodulin-dependent protein kinase type II subunit alpha; Calcium/calmodulin-dependent protein kinase type II subunit beta; Calcium/calmodulin-dependent protein kinase type II subunit delta; CaM kinase II alpha subunit; CaM kinase II beta subunit; CaM kinase II delta subunit; CaM kinase II subunit alpha; CaM kinase II subunit beta; CaM kinase II subunit delta; CaM-kinase II alpha chain; CaM-kinase II beta chain; CaM-kinase II delta chain; CAMK; CaMK II; CaMK-2 delta; CaMK-2a; CAMK-II; CaMK-II alpha subunit; CaMK-II delta subunit; caMK-II subunit alpha; caMK-II subunit beta; caMK-II subunit delta; CaMK2 delta; CaMK2- delta; CaMK2-a; CAMK2G; CAMKG; CaMKIINalpha; proline rich calmodulin-dependent protein kinase
Gene Aliases: 2810011D23Rik; 8030469K03Rik; [d]-CaMKII; CAM2; CaMK II; CAMK2; CAMK2A; CAMK2B; CAMK2D; CAMKA; CAMKB; CAMKD; CaMKII; KIAA0968; Kiaa4163; mKIAA0968; R74975
UniProt ID: (Human) Q9UQM7, (Human) Q13554, (Human) Q13557, (Mouse) P11798, (Mouse) P28652, (Mouse) Q6PHZ2
Entrez Gene ID: (Human) 815, (Human) 816, (Human) 817, (Mouse) 12322, (Mouse) 12323, (Mouse) 108058
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support