Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | View 5 publications 5 publications |
Immunohistochemistry (IHC) |
- | View 2 publications 2 publications |
Immunocytochemistry (ICC/IF) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Chicken, Human, Mouse, Rabbit, Rat, Xenopus |
Published species |
Human, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human ETS1 that contains threonine 38. |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
Antigen affinity chromatography |
Storage buffer |
Dulbecco's PBS, pH 7.3, with 1mg/mL BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2533574 |
ETS1 (c-Ets-1; v-ets erythroblastosis virus E26 oncogene homlog 1 (avian); ETS-1; EWSR2; p54), a 441 A. A protein, is a member of DNA-binding ETS protein family. It is a multifunctional protein consisting of an N-terminal pointed domain, a central transactivation domain and 2 auto inhibitory domains flanked on both sides of conserved C-terminal Ets domain. It interacts with factors including PEBP2alphaA, p300/CBP and mediates transactivation of Osteopontin, Presenilin-1, Stromelysin and Collagenase. It is activated by Ras/MAPK signaling and its expression is up-regulated in resting T-cells. Its activity is modulated by various factors including AP-1, Sp-1, c-myb and MafB. Sp100 activates ETS1 in nuclear bodies. It interacts with EAP1/Daxx and causes repression of MMP1 and BCL2 transactivation. It interacts with STAT6 and modulates Socs-1 expression induced by IL-4. It interacts with GATA3 and positively regulates Tax-1 dependent IL-5 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1; avian erythroblastosis virus E26 oncogene 1; c-ets protein; c-ets-1 oncogene; c-ets-1 protein; c-ets-alpha; Ets avian erythroblastosis virus E2 oncogene homolog 1 (tumor progression locus 1); ets protein; Ets-1Delta(III-VI); FLJ10768; oncoprotein; p54; p54 transcription factor; Protein C-ets-1; TNFAIP3 interacting protein 1; Transforming protein p54/c-ets-1; transforming protein p68/c-ets-1; tumor progression locus 1; unnamed protein product; v-ets avian erythroblastosis virus E2 oncogene homolog 1; v-ets avian erythroblastosis virus E26 oncogene homolog 1; v-ets erythroblastosis virus E26 oncogene homolog 1; virion-associated nuclear-shuttling protein
Gene Aliases: AI196000; AI448617; c-ets-1; c-ets-beta; c-ets1; D230050P06; ETS-1; ETS-1A; ETS-1B; ETS1; Etsoncb; EWSR2; p54; TNIP1; Tpl1
UniProt ID: (Chicken) P13474, (Human) P14921, (Rat) P41156, (Mouse) P27577
Entrez Gene ID: (Chicken) 396235, (Human) 2113, (Rabbit) 100009373, (Rat) 24356, (Mouse) 23871
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support