Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Regulates phosphorylation of a number of proteins involved in translation regulation including EIF2A, EIF4EBP1 and RPS6KB1. May be involved in the endoplasmic reticulum stress response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 39 protein; family with sequence similarity 129, member A; Protein FAM129A; Protein Niban 1
Gene Aliases: C1orf24; FAM129A; GIG39; NIBAN; NIBAN1
UniProt ID: (Human) Q9BZQ8
Entrez Gene ID: (Human) 116496
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support