Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of FBP1 only when phosphorylated at Tyr265.
Fructose-1,6-bisphosphatase 1, a gluconeogenesis regulatory enzyme, catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. Fructose-1,6-diphosphatase deficiency is associated with hypoglycemia and metabolic acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; EC 3.1.3.11; FBP 1; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; fructose-1; fructose-1,6- biphosphatase 1; Fructose-1,6-bisphosphatase 1; Fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; growth-inhibiting protein 17; Liver FBPase
Gene Aliases: FBP; Fbp-2; FBP1; Fbp2; Fbp3; Fdp
UniProt ID: (Human) P09467, (Rat) P19112, (Mouse) Q9QXD6
Entrez Gene ID: (Human) 2203, (Rat) 24362, (Mouse) 14121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support