Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide sequence around phosphorylation site of Serine 401(K-M-S(p)-N-K) derived from Human GATA2. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2554191 |
A suggested positive control for Western blot is JK cells.
GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial transcription factor GATA-2; FLJ45948; GATA-binding protein 2; MGC2306
Gene Aliases: DCML; Gata-2; GATA2; IMD21; MONOMAC; NFE1B
UniProt ID: (Human) P23769, (Mouse) O09100
Entrez Gene ID: (Human) 2624, (Mouse) 14461
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support