Invitrogen
FIGURE: 1 / 2
MA1-24751 detects Histone H3 (phospho S28) from human, mouse, bovine, and hamster samples.
MA1-24751 has been successfully used in immunocytochemistry, flow cytometry and Western blot procedures. Western blot detects a band at ~15 kDa. A suggested positive control is whole extract of cultured human acute T cell leukemia Jurdat cells, treated with Nocodazole.
The MA1-24751 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 23-35 of human Histone H3.
Store at 4ºC short term. For long term storage, store at -20ºC, avoiding freeze/thaw cycles.
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.