Search Thermo Fisher Scientific
FIGURE: 1 / 1
PA1-28418 detects MEK7 from human samples.
PA1-28418 has been successfully used in Western blot applications.
The PA1-28418 immunogen is: Synthetic peptide (Human) derived from a region of human MEK 7 that contains serine 271 and threonine 275. The sequence is conserved in mouse.
The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase family. This kinase specifically activates MAPK8/JNK1 and MAPK9/JNK2, and this kinase itself is phosphorylated and activated by MAP kinase kinases including MAP3K1/MEKK1, MAP3K2/MEKK2, MAP3K3/MEKK5, and MAP4K2/GCK. This kinase is involved in the signal transduction mediating the cell responses to proinflammatory cytokines, and environmental stresses. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found, but only one transcript variant has been supported and defined.