Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 565 kDa.
Purity is >95% by SDS-PAGE.
Dihydropyridine receptor (DHPR) is a surface membrane protein critical for the excitation-contraction coupling of striated muscle. DHPR and the sarcoplasmic reticulum ryanodine receptor (RyR) are two key components of the intracellular junctions, where depolarization of the surface membrane is converted into the release of Ca2+ from internal stores. The alpha1-subunit of the DHPR contains a cytoplasmic loop which is thought to be involved in the interactions with RyR. Phosphorylation of the DHPR alpha1-subunit is also thought to play a role in the functional interaction of DHPR and RyR. Mutation in DHPR alpha1 results in excitation-contraction uncoupling, leading to muscular dysgenesis, a complete inactivity in developing skeletal muscles. Cells that do not express RyR also lack excitation-contraction coupling and exhibit a severalfold reduction in Ca2+ current density.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium release channel; Cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac-type ryanodine receptor; hRYR-2; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor type 2; ryanodine receptor type II; RYR-2; Type 2 ryanodine receptor
Gene Aliases: 9330127I20Rik; ARVC2; ARVD2; RyR; RYR-2; RYR2; VTSIP
UniProt ID: (Human) Q92736, (Rat) B0LPN4, (Mouse) E9Q401
Entrez Gene ID: (Human) 6262, (Rat) 689560, (Mouse) 20191
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support