Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
A suggested positive control for Western blot is HeLa or A549 cells; suggested positive control for IHC is human brain tissue.
This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90 kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC104792; MGC34446; proliferation-inducing gene 49; Protein Tob1; Transducer of erbB-2 1; transducer of erbB2; transducer of ERBB2, 1
Gene Aliases: APRO5; APRO6; PIG49; TOB; TOB1; TROB; TROB1
UniProt ID: (Human) P50616, (Mouse) Q61471
Entrez Gene ID: (Human) 10140, (Mouse) 22057, (Rat) 170842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support