Product References
Imatinib as a key inhibitor of the platelet-derived growth factor receptor mediated expression of cell surface heparan sulfate proteoglycans and functional properties of breast cancer cells.
The FEBS journal
Malavaki CJ,Roussidis AE,Gialeli C,Kletsas D,Tsegenidis T,Theocharis AD,Tzanakakis GN,Karamanos NK
MA1-12439 was used in western blot to study the effects of Glivec on breast cancer heparin sulfate proteogycan expression and cell proliferation
Wed May 01 00:00:00 EDT 2013
RhoH plays critical roles in Fc epsilon RI-dependent signal transduction in mast cells.
Journal of immunology (Baltimore, Md. : 1950)
Oda H,Fujimoto M,Patrick MS,Chida D,Sato Y,Azuma Y,Aoki H,Abe T,Suzuki H,Shirai M
MA1-12439 was used in western blot to study the role of RhoH in Fc epsilon RI-dependent signal transduction in mast cells
Thu Jan 15 00:00:00 EST 2009
Functional implications of altered subcellular localization of PELP1 in breast cancer cells.
Cancer research
Vadlamudi RK,Manavathi B,Balasenthil S,Nair SS,Yang Z,Sahin AA,Kumar R
MA1-12439 was used in immunoprecipitation to investigate the role of altered subcellular localization of PELP1 in breast cancer cells
Thu Sep 01 00:00:00 EDT 2005
The combi-targeting concept: dissection of the binary mechanism of action of the combi-triazene SMA41 in vitro and antitumor activity in vivo.
The Journal of pharmacology and experimental therapeutics
Matheson SL,McNamee JP,Wang T,Alaoui-Jamali MA,Tari AM,Jean-Claude BJ
MA1-12439 was used in western blot to study the anti-tumor mechanism of a unimolecular combination of an EGFR tyrosine kinase inhibitor and a DNA damaging triazine
Wed Dec 01 00:00:00 EST 2004
Delivery of insulin-like growth factor-I to the rat brain and spinal cord along olfactory and trigeminal pathways following intranasal administration.
Neuroscience
Thorne RG,Pronk GJ,Padmanabhan V,Frey WH
MA1-12439 was used in immunohistochemistry to evaluate a specific method for IGF-I delivery into CNS system through intranasal administration
Tue Nov 09 00:00:00 EST 2004
Correlation of P-cadherin and beta-catenin expression and phosphorylation with carcinogenesis in rat tongue cancer induced with 4-nitroquinoline 1-oxide.
Oral oncology
Tamura I,Sakaki T,Chaqour B,Howard PS,Ikeo T,Macarak EJ
MA1-12439 was used in immunohistochemistry and western blot to study the role of P-cadherin and beta-catenin in 4-nitroquiniline-1-oxide-induced tongue cancer tumorigenesis
Tue Jul 01 00:00:00 EDT 2003
Inhibition of epidermal growth factor receptor-mediated signaling by "Combi-triazene" BJ2000, a new probe for Combi-Targeting postulates.
The Journal of pharmacology and experimental therapeutics
Brahimi F,Matheson SL,Dudouit F,McNamee JP,Tari AM,Jean-Claude BJ
MA1-12439 was used in western blot to study the inhibition of EGFR-mediated signaling by a "Combi-triazene" molecule with binary EGFR targeting/DNA-damaging properties
Tue Oct 01 00:00:00 EDT 2002
Dual action of estrogen on glutamate-induced calcium signaling: mechanisms requiring interaction between estrogen receptors and src/mitogen activated protein kinase pathway.
Brain research
Nilsen J,Chen S,Brinton RD
MA1-12439 was used in immunocytochemistry to investigate the mechanism for the effect of estrogen on glutamate-induced calcium signaling
Fri Mar 15 00:00:00 EST 2002
Intracellular distribution of lysosomal sialidase is controlled by the internalization signal in its cytoplasmic tail.
The Journal of biological chemistry
Lukong KE,Seyrantepe V,Landry K,Trudel S,Ahmad A,Gahl WA,Lefrancois S,Morales CR,Pshezhetsky AV
MA1-12439 was used in immunoprecipitation to study the role of tyrosine phosphorylation of the cytoplasmic internalization signal of lysosomal sialidase in regulating its subcellular localization
Fri Dec 07 00:00:00 EST 2001
Alloantibody-mediated class I signal transduction in endothelial cells and smooth muscle cells: enhancement by IFN-gamma and TNF-alpha.
Journal of immunology (Baltimore, Md. : 1950)
Bian H,Reed EF
MA1-12439 was used in western blot to test if anti-HLA Abs transduce signals via HLA class I molecules and stimulate cell proliferation.
Thu Jul 15 00:00:00 EDT 1999
Ligation of HLA class I molecules on smooth muscle cells with anti-HLA antibodies induces tyrosine phosphorylation, fibroblast growth factor receptor expression and cell proliferation.
International immunology
Bian H,Harris PE,Reed EF
MA1-12439 was used in western blot to elucidate the role of anti-donor HLA antibodies in transplant atherosclerosis.
Tue Sep 01 00:00:00 EDT 1998