Search Thermo Fisher Scientific
PG 5.1 represents an excellent marker for all forms of intercellular adhering junctions, such as: desmosomes of epithelial and myocardial cells (incl. cultured cells); zonulae and fasciae adherentes of epithelia, endothelia of blood vessels and myocardial cells; adherens-type junctions (e.g. lens tissue, pigmented retinal cells, Sertoli cells of testis). The PG 5.1 epitope maps within the C-terminus at the extreme end of repeat 13 (aa 632-687) of plakoglobin. Polypeptide reacting: MW 83 000 Plakoglobin, "band 5" polypeptide of intercellular adhering junctions (identical to gamma-catenin)Reactivity on cultured cell lines: Cell cultures forming monolayers .
JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.