Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDGELSCRVF QLISLMDTGL PRCCNEKIEL AILWFLDQFR KTYVGDQLQR TSKVYARMSE VLGITDDNHV LETFMT
Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The transport of protein and large RNAs through the nuclear pore complexes (NPC) is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal (NLS) is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN (MIM 601179) plays a key role in NLS-dependent protein import. RAN-binding protein-17 is a member of the importin-beta superfamily of nuclear transport receptors.[supplied by OMIM, Jul 2002].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ran-binding protein 17
Gene Aliases: RANBP17
UniProt ID: (Human) Q9H2T7
Entrez Gene ID: (Human) 64901
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support