Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (C)ces type 1 Rhesus blood group D antigen; Blood group Rh(CE) polypeptide; blood group RhCcEe antigen; blood group RhCE polypeptide; CD240CE; Rh blood group antigen Evans; Rh blood group C antigen; Rh blood group protein; Rh blood group, CcEe antigens; Rh polypeptide 1; Rh polypeptide I; Rh30A; RhCE blood group antigens; RHCE blood group variant Crawford antigen Rh43; rhesus blood group antigen, RhC antigen; Rhesus blood group CE protein; Rhesus blood group E antigen; rhesus blood group little e antigen; Rhesus blood group Rhce antigen; Rhesus C/E antigens; Rhesus system C and E polypeptides; RhIXB; RhPI; silenced Rh blood group CcEe antigen; truncated RhCcEe antigen; truncated RHCE
Gene Aliases: CD240CE; RH; RH30A; Rh4; RHC; RHCE; RHE; RhIVb(J); RHIXB; RHPI; RhVI; RhVIII
UniProt ID: (Human) P18577
Entrez Gene ID: (Human) 6006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support