Search Thermo Fisher Scientific
Recommended positive controls: U87-MG, mouse ovary.
Predicted reactivity: Mouse (97%), Xenopus laevis (80%), Pig (97%), Chicken (82%), Sheep (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plasma membrane-associated small GTPase which cycles between an active GTP-bound and an inactive GDP-bound state. Causes the formation of thin, actin-rich surface projections called filopodia. Functions cooperatively with CDC42 and Rac to generate additional structures, increasing the diversity of actin-based morphology.