Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAHITINQYL QQVYEAIDSR DGASCAELVS FKHPHVANPR LQMASPEEKC QQVLEPPYDE MFAAHLRCTY AVGNHDFIEA YKCQTVIVQS FLRAFQAHKE ENWALPVMYA VALDLRVFAN NADQQLVKKG KSKVGDMLEK AAELLMSCFR VCASDTRAGI EDSKKWGMLF LVNQLLKIYF KINKLHLCKP LIRAIDSSNL KDDYSTAQRV TYKYYVGRKA MFDSDFKQAE EYLSFAFEHC HRSSQKNKRM ILIYLLPVKM LLGHMPTVEL LKKYHLMQFA EVTRAVSEGN LLLLHEALAK HEAFFIRCGI FLILEKLKII TYRNLFKKVY LLLKTHQLSL DAFLVALKFM QVEDVDIDEV QCILANLIYM GHVKGYVSHQ HQKLVVSKQN PFPPLSTVC
Required for B-cell survival through the regulation of the expression of cell-cycle checkpoint MAD2L1 protein during B cell differentiation. Component of the TREX-2 complex (transcription and export complex 2), composed of at least ENY2, GANP, PCID2, DSS1, and either centrin CETN2 or CETN3 (PubMed:22307388). The TREX-2 complex functions in docking export-competent ribonucleoprotein particles (mRNPs) to the nuclear entrance of the nuclear pore complex (nuclear basket). TREX-2 participates in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery. Binds and stabilizes BRCA2 and is thus involved in the control of R-loop-associated DNA damage and transcription-associated genomic instability. R-loop accumulation does not increase in PCID2-depleted cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSN12-like protein; PCI domain-containing protein 2; RP11-98F14.6
Gene Aliases: F10; HT004; PCID2
UniProt ID: (Human) Q5JVF3
Entrez Gene ID: (Human) 55795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support