Search Thermo Fisher Scientific
MA1-90620 detects RPA34 from human, hamster samples.
MA1-90620 has been successfully used in immunofluorescence, immunohistochemistry (paraffin tissue), immunoprecipitation, Western blot applications.
The MA1-90620 immunogen is: Tissue / cell preparation (Human); purified from U293 cells.
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.