Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Isotype Controls ›
          • Rat IgG2a Isotype Controls
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productDetailViewModel.formattedProductSubType.equals('ELISA') ? 'ELISA kits' : 'antibodies'}} or contact Technical Support with any questions.

          Invitrogen

          Rat IgG2a Isotype Control, PE

          View all Rat IgG2a Isotype Controls

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Datasheet
          Certificates
          Datasheet
          Certificates

          Cite Rat IgG2a Isotype Control, PE

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          PA5-33190

          Applications
          Tested Dilution

          Flow Cytometry (Flow)

          Assay-dependent

          Control (Ctrl)

          Assay-Dependent
          Product Specifications

          Host/Isotype

          Rat / IgG2a

          Class

          Control

          Type

          Isotype Control

          Immunogen

          Ovalbumin - DNP

          Conjugate

          PE PE PE

          Excitation/Emission Max

          565/576 nm View spectra spectra

          Form

          Lyophilized

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Wet ice

          Product Specific Information

          PA5-33190 is suitable for use as a negative control for the measurement of non-specific binding of rat monoclonal antibodies of isotype IgG2a in human tissues. Test results show that PA5-33190 is also suitable for use as a negative control with canine tissues.

          Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4°C in the dark. Once reconstituted, this product has a shelf life of approx. 18 months.

          For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.

          Target Information

          Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody PA5-33190 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline