Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-06810 detects reticulon-1A (NSP-A) in human, mouse, hamster, and rat samples.
MA1-06810 has sucessfully been used in Western blotting, FACS, ICC and immunohistochemistry procedures. By Western blot it detects a ~135 kDa band representing reticulon-1A.
The MA1-06810 immunogen is a partially purified bacterially expressed reticulon-1A (NSP-A) hybrid protein.
Reticulon-1 was formerly designated NSP for Neuroendocrine-Specific-Protein, because it is specifically expressed in neural and neuroendocrine tissues. The NSP-gene has been mapped by fluorescence in situ hybridization to human chromosome 14q21-q22. The NSP-gene encodes three overlapping proteins, i.e. reticulon-1A (NSP-A), reticulon-1B (NSP-B), and reticulon-1C (NSP-C). These proteins were found to be anchored to membranes of the endoplasmic reticulum through their common carboxy-terminal regions. Reticulon-1A is a protein with a molecular weight of approximately 135 kDa, which occurs in various isoforms presumably depending on the degree of phosphorylation of serine residues. In lung cancer diagnosis Reticulon-1A appeared to be a reliable marker for the detection of neuroendocrine differentiation, since most of the small cell lung carcinoma (SCLC) and carcinoid tumors showed expression of Reticulon-1A. Reticulon-1B is a phosphoprotein with a MW of 45 kDa and is restricted to the lung cancer cell line NCI-H82. Reticulon-1B is so far not found in human tissues. Reticulon-1C is a protein with a MW of 23 kDa which is not phosphorylated and is found with Reticulon-1A in SCLC (cell lines) and not in non-SCLC (cell cultures).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 0710005K15Rik; 4930441F12Rik; MGC133250; MGC40985; Neuroendocrine-specific protein; R75279; reticulon 1; Reticulon-1; RP23-450G14.1; Rtn1-a; Rtn1-b; Rtn1-c; S-rex
Gene Aliases: I79_024631; NSP; RTN1
UniProt ID: (Human) Q16799, (Rat) Q64548, (Mouse) Q8K0T0
Entrez Gene ID: (Chinese hamster) 100750648, (Human) 6252, (Rat) 116644, (Mouse) 104001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support