Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 14
MA1-123 detects GTPases RhoA and RhoC, but does not react with RhoB.
MA1-123 has been successfully used in Western blot, immunofluorescence, IHC (P) and immunoprecipitation applications with human and mouse samples.
RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aplysia ras-related homolog 12; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; h12; h9; oncogene RHO H12; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 9; Rho family GTPase; Rho-related GTP-binding protein RhoC; rhoC GTPase; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; Transforming protein RhoA
Gene Aliases: AI324259; ARH12; ARH9; ARHA; Arha1; Arha2; ARHC; H9; RHO12; RHOA; RHOC; RHOH12; RHOH9
UniProt ID: (Human) P61586, (Human) P08134, (Rat) P61589, (Mouse) Q9QUI0, (Mouse) Q62159
Entrez Gene ID: (Human) 387, (Human) 389, (Rat) 117273, (Mouse) 11848, (Mouse) 11853, (Rat) 295342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support