Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
S100A14 gene encodes a member of the S100 protein family which contains an EF-hand motif and binds calcium. The gene is located in a cluster of S100 genes on chromosome 1. Levels of the encoded protein have been found to be lower in cancerous tissue and associated with metastasis suggesting a tumor suppressor function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: breast cancer membrane protein 84; Protein S100-A14; S100 calcium-binding protein A14; S114
Gene Aliases: BCMP84; S100A14; S100A15
UniProt ID: (Human) Q9HCY8
Entrez Gene ID: (Human) 57402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support