Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 48 kDa protein; arrestin 1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; Rod photoreceptor arrestin; S-AG; S-antigen; S-antigen; retina and pineal gland (arrestin); S-arrestin; visual arrestin 1
Gene Aliases: A930001K18Rik; Arr1; arrestin; Irbp; RP47; S-AG; SAG; SAGMR; SANTI
UniProt ID: (Human) P10523, (Rat) P15887, (Mouse) P20443
Entrez Gene ID: (Human) 6295, (Rat) 25539, (Mouse) 20215
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support