Search Thermo Fisher Scientific
PA1-31762 detects SAP130 from human samples.
PA1-31762 has been successfully used in Western blot applications.
The PA1-31762 immunogen is: Peptide with sequence VSKKLEDIRTRYAF, from C terminus of the protein sequence according to NP_036558.
SAP130 is a subunit of the histone deacetylase-dependent SIN3A corepressor complex (Fleischer et al., 2003).